Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
Plug cable combination, K 2 S HT (coaxial electrode plug, 2 m coaxial cable, 135 °C, electrode socket (extension cable)), 2 m
Catalog # 662-2208
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2208
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
Plug cable combination, K 2 S HT (coaxial electrode plug, 2 m coaxial cable, 135 °C, electrode socket (extension cable)), 2 m
Price
120,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Plug cable combination, K 2 S HT (coaxial electrode plug, 2 m coaxial cable, 135 °C, electrode socket (extension cable))
  • Length:
    2 m
  • Seq:
    14