Receiving flasks
1000 ml, Low temperature, KS 35/20
BUCH040775
: BUCHI
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
- Socket:KS 35/20
- Type:Low temperature
- Capacity:1000 ml