ContactUsLinkComponent
KIT METAL 2,8MM IN 2 ML TUBES 50 PREP, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 2,8mm stainl ess steel beads in 2mL standard tubes,for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, c 1 * 1 KIT
Catalog # BERT03961-1-001
Supplier:  Bertin Instruments
KIT METAL 2,8MM IN 2 ML TUBES 50 PREP, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 2,8mm stainl ess steel beads in 2mL standard tubes,for hard tissue grinding like muscle, skin, spincor, bone, teeth, hair, seeds, c 1 * 1 KIT
Catalog # BERT03961-1-001
Supplier:  Bertin Instruments
Frequently Bought Together