Orders LinkContactUsLinkComponent
Objectives
EC-Planachromatic Neofluar, M27, N.A. of 1,3, WD of 0,20 mm
Catalog # ZEIS4204909900000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4204909900000
Supplier:  ZEISS
Objectives
Objectives
EC-Planachromatic Neofluar, M27, N.A. of 1,3, WD of 0,20 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    100×
  • Description:
    EC-Planachromatic Neofluar, M27, N.A. of 1,3, WD of 0,20 mm
  • Seq:
    23