Cannulas for micro syringes, Dosys™
Dispensing cannula, Stainless steel, 2,2×100 mm
SOCO370.22100
Blunt end cannulas provide precise distribution with Luer nozzle microdispensers.
- Ø×L:2,2×100 mm
- Description:Dispensing cannula
- Material:Stainless steel
- Seq:002