Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Cover Slips, Rectangular

VWR®, Cover Slips, Rectangular

24×32 mm, #1
  631-1572
VWR®, Cover Slips, Rectangular
  631-1572
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular

24×32 mm, #1

 
61,80 €
   
1 * 1.000 ST
 

These cover slips are specially designed for diagnostic applications.

  • Made of pure white Hydrolytic Class 1 glass

Thickness: 0.13 - 0.16 mm

Thickness: 0,13 - 0,16 mm

Packaging: In dispenser boxes with separate lid.

 
  • No.:
    #1
  • L×W:
    24×32 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.