Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Cover Slips, Rectangular

VWR®, Cover Slips, Rectangular

21×26 mm, #1
Catalog # 631-1569
Supplier:   VWR Collection
VWR®, Cover Slips, Rectangular
Catalog # 631-1569
Supplier:  VWR Collection
VWR®, Cover Slips, Rectangular
VWR®, Cover Slips, Rectangular

21×26 mm, #1

Price
106,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents

These cover slips are specially designed for diagnostic applications.

  • Made of pure white Hydrolytic Class 1 glass

Thickness: 0.13 - 0.16 mm

Thickness: 0,13 - 0,16 mm

 : In dispenser boxes with separate lid.

Specifications
  • No.:
    #1
  • L×W:
    21×26 mm
  • Seq:
    06