Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR®, Microscope Slides

Slide with cut edges and plain end, Transparent, Float glass, 1,0 mm, 76×26 mm
  631-1550
VWR®, Microscope Slides
  631-1550
VWR®, Microscope Slides
VWR®, Microscope Slides

Slide with cut edges and plain end, Transparent, Float glass, 1,0 mm, 76×26 mm

 
5,50 €
   
1 * 50 ST
 
  • Colour:
    Transparent
  • Description:
    Slide with cut edges and plain end
  • L×W:
    76×26 mm
  • Material:
    Float glass
  • Seq:
    01
  • Thickness:
    1,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.