Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bath fluids

DW-Therm HT®, 20...340 °C, ~190 °C, 32 mm²/s, 10 L
Catalog # HUBE6673
Supplier:   HUBER
Bath fluids
Catalog # HUBE6673
Supplier:  HUBER
CAS Number:  
Bath fluids
Bath fluids

DW-Therm HT®, 20...340 °C, ~190 °C, 32 mm²/s, 10 L

CAS Number:  
Price
725,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.

All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.

Caution: * –20...+235 °C for closed system applications

Use biocides safely. Always read the label and product information before use

Specifications

Specifications

  • Pk:
    10 L
  • Flash point:
    ~190 °C
  • Temperature range:
    20...340 °C
  • Type:
    DW-Therm HT®
  • Viscosity:
    32 mm²/s

Specifications