Filter pump
Water pump 5751, with non return valve connector plug, G 1/2 (199 g)
Catalog # USBE5751
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Water jet pump in nickel-plated brass.
- Stable and solid
- A side knob allows connection to a vacuum pipe with an interior diameter of 8 mm
- Available with or without non-return valve
- Supplied with water connection for pipes with an internal diameter of 14 mm or bolting for connection to a tap
- Pressure gauge available as an option for all models
Documents
USBE5721_TDS_EN
Technisches Datenblatt
Specifications
- Description:Water pump 5751, with non return valve connector plug, G 1/2 (199 g)
Accessories
Product Information
- Catalog #AvailabilityPrice
- Water pump manometer
Product Information
- Catalog #AvailabilityPrice
- G 1/2 connection for water pump