BioChemi HR camera: 12 bit monochrome, 16 File Bit Depth (A/D), digital camera and built-in camera bracket, 2048 x 2048 chip with binning capability (4Mp). Two stage, forced air assisted, peltier cooling to - 45C from ambient, MANUAL fixed f/0.95 le 1 * 1 items
Catalog # 733-2023
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together