Orders LinkContactUsLinkComponent
Boxes, with Hinged Lid
Boxes, with Hinged Lid
without label holder, 185×140×80 mm
Catalog # SEMA0341
Supplier:   SEMADENI
Boxes, with Hinged Lid
Catalog # SEMA0341
Supplier:  SEMADENI
Boxes, with Hinged Lid
Boxes, with Hinged Lid
without label holder, 185×140×80 mm
Price
13,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
The practical hinged flip-top box container is made from dimensionally stable Polypropylene. The standard colour is grey and the range includes a total of five different sizes. With lid and container made from a single block, snap-on cap and integral hinge, you can package your small item safely and securely.
  • Heavyweight boxes
  • Rigid and robust
  • Stackable
  • PP, grey
Specifications
  • Type:
    without label holder
  • W×D×H:
    185×140×80 mm