Multiscreen® SEQ₃₈₄ Filter Plates
Multiscreen® SEQ₃₈₄ filter plate, Clear, 100 µl, 384, -
MILFS384SEQ10
: Merck
The MultiScreen384-SEQ filter plate is suited for use with most liquid handling systems. The filter plate operates by vacuum filtration enabling top-loading and recovery of sample. Our plate is designed with the needs of the high throughput laboratory in mind, meets SBS standards and is ideally suited for 'walk away' automation.
- Automation friendly: Design and protocol saves time, money and improves reproducibility
- Efficient dye terminator & salt removal: Recovers highly purified sequencing products for long, accurate sequencing reads
- Eliminates dry-down step: Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
- Saves reagents: plates suitable for use with ¹/₁₆ sequencing reactions
- Proprietary membrane
- Color code: Clear
- Automation compatible: No
Application: Research category, assays, general filtration and nucleic acid.
Packaging: Does not include reagents.
- No. of wells:384
- Recommended working volume:100 µl
- Sterile:-
- Colour:Clear
- Description:Multiscreen® SEQ₃₈₄ filter plate
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



