Orders LinkContactUsLinkComponent
Funnels
Funnels
200 mm
Catalog # 221-0165
Supplier:   NICKEL ELECTRO
Funnels
Catalog # 221-0165
Supplier:  NICKEL ELECTRO
Funnels
Funnels
200 mm
Price
51,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Good chemical resistance, extremely high thermal resistance
  • With air duct for discharging container air
  • Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Specifications
  • Funnel O-Ø:
    200 mm
  • Seq:
    03