Orders LinkContactUsLinkComponent
Funnels
Funnels
150 mm
Catalog # NICKFUNSS150H
Supplier:   NICKEL ELECTRO
Funnels
Catalog # NICKFUNSS150H
Supplier:  NICKEL ELECTRO
Funnels
Funnels
150 mm
Price
37,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Good chemical resistance, extremely high thermal resistance
  • With air duct for discharging container air
  • Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Specifications
  • Funnel O-Ø:
    150 mm
  • Seq:
    02