Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Standard, 2 - 300 µl, -, Transparent, Gilson®, Sartorius, Finnpipette®, Labsystem®, Rack
Catalog # KART9644
Supplier:   KARTELL
Pipette tips
Catalog # KART9644
Supplier:  KARTELL
Pipette tips
Pipette tips
Standard, 2 - 300 µl, -, Transparent, Gilson®, Sartorius, Finnpipette®, Labsystem®, Rack
Price
157,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips are autoclavable to +121 °C for 20 minutes
Specifications
  • Packed:
    Rack
  • Sterile:
    -
  • Volume:
    2 - 300 µl
  • Seq:
    15
  • Tip style:
    Standard
  • Tip colour:
    Transparent
  • For use with:
    Gilson®, Sartorius, Finnpipette®, Labsystem®