Order Entry
Austria
Orders LinkContactUsLinkComponent
Accessories for RapidVap®

Accessories for RapidVap®

PTFE-coated aluminium block for 110 tubes Ø 16 mm, max. height 150 mm
Catalog # 531-0208
Supplier:   LABCONCO
Accessories for RapidVap®
Catalog # 531-0208
Supplier:  LABCONCO
Accessories for RapidVap®
Accessories for RapidVap®

PTFE-coated aluminium block for 110 tubes Ø 16 mm, max. height 150 mm

Price
2 700,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    PTFE-coated aluminium block for 110 tubes Ø 16 mm, max. height 150 mm
  • Seq:
    07