Filter funnels
1000 ml, 120 mm, 3
LENZ07195370
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
- Plate Ø:120 mm
- Porosity:3
- Capacity:1000 ml



