Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter funnels

Filter funnels

1000 ml, 120 mm, 2
Catalog # LENZ07195270
Supplier:   Lenz Laborglas
Filter funnels
Catalog # LENZ07195270
Supplier:  Lenz Laborglas
Filter funnels
Filter funnels

1000 ml, 120 mm, 2

Price
145,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With integrated sintered glass filters disc.

Documents

Specifications
  • Plate Ø:
    120 mm
  • Porosity:
    2
  • Capacity:
    1000 ml