Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter funnels

Filter funnels

50 ml, 35 mm, 2
Catalog # LENZ07195228
Supplier:   Lenz Laborglas
Filter funnels
Catalog # LENZ07195228
Supplier:  Lenz Laborglas
Filter funnels
Filter funnels

50 ml, 35 mm, 2

Price
30,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With integrated sintered glass filters disc.

Documents

Specifications
  • Plate Ø:
    35 mm
  • Porosity:
    2
  • Capacity:
    50 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.