Filter discs
2, 50 mm
Catalog # LENZ07180250
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Glass filter discs centred with unfused rim.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Ø:50 mm
- Porosity:2
- Seq:17