Order Entry
Austria
Orders LinkContactUsLinkComponent
Viscous samplers

Viscous samplers

Viscous sampler for medium viscosity materials, Stainless steel 316, 300 ml, 1000 mm
Catalog # 231-0487
Supplier:   Sampling Systems
Viscous samplers
Catalog # 231-0487
Supplier:  Sampling Systems
Viscous samplers
Viscous samplers

Viscous sampler for medium viscosity materials, Stainless steel 316, 300 ml, 1000 mm

Price
1 010,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These viscous samplers are designed for creams, shampoos, soaps, honey and other semi-viscous materials.

After sampling, the sampler can be completely dismantled, for a quick and thorough cleaning.

Specifications
  • Capacity:
    300 ml
  • Description:
    Viscous sampler for medium viscosity materials
  • Length:
    1000 mm
  • Material:
    Stainless steel 316
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.