Orders LinkContactUsLinkComponent
Density bottles, adjusted
Density bottles, adjusted
25* ml
Catalog # LENZ05630222
Supplier:   Lenz Laborglas
Density bottles, adjusted
Catalog # LENZ05630222
Supplier:  Lenz Laborglas
Density bottles, adjusted
Density bottles, adjusted
25* ml
Price
72,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
  • The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
  • The top of stopper is ground and polished for ease of wiping clean
  • Stopper is size NS 10/19 with capillary
* DIN ISO 3507
Documents
Specifications
  • Volume (nom.):
    25* ml