Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Extraction insert for solids, for extractor 100 ml capacity (250 ml volume)
Catalog # LENZ05431202
Supplier:   Lenz Laborglas
Extraction thimbles
Catalog # LENZ05431202
Supplier:  Lenz Laborglas
Extraction thimbles
Extraction thimbles
Extraction insert for solids, for extractor 100 ml capacity (250 ml volume)
Price
41,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass insert with bores for solids.
Dokumente
Specifications
  • Description:
    Extraction insert for solids, for extractor 100 ml capacity (250 ml volume)
  • Seq:
    11