Orders LinkContactUsLinkComponent
Threaded ball flanges
Threaded ball flanges
Socket KF 15 with steel thread M 16×1
Catalog # 201-1163
Supplier:   Lenz Laborglas
Threaded ball flanges
Catalog # 201-1163
Supplier:  Lenz Laborglas
Threaded ball flanges
Threaded ball flanges
Socket KF 15 with steel thread M 16×1
Price
124,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of stainless steel.
  • Very good chemical resistance
  • High temperature resistance
  • Sturdy
With KF socket, with thread.
Specifications
  • Type:
    Socket KF 15 with steel thread M 16×1