Orders LinkContactUsLinkComponent
Stoppers
Stoppers
14/23, 12,2 mm
Catalog # KART1455
Supplier:   KARTELL
Stoppers
Catalog # KART1455
Supplier:  KARTELL
Stoppers
Stoppers
14/23, 12,2 mm
Price
37,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Ø bottom:
    12,2 mm
  • NS:
    14/23