Order Entry
Austria
Orders LinkContactUsLinkComponent
Multi-channel pipettes, Multipet

Multi-channel pipettes, Multipet

50 - 300 µl, ±1,6 (50 µl); ±1,0 (300 µl) %, ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
Catalog # KART1328.10
Supplier:   KARTELL
Multi-channel pipettes, Multipet
Catalog # KART1328.10
Supplier:  KARTELL
Multi-channel pipettes, Multipet
Multi-channel pipettes, Multipet

50 - 300 µl, ±1,6 (50 µl); ±1,0 (300 µl) %, ≤ ±3,0 (50 µl); ±1,0 (300 µl) %

Price
1 740,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These multi-channel micropipettes with air cushion are ideal for dispensing in multiwell plates for ELISA dosages, molecular biology and DNA amplifications.

  • Volume can be easily selected by turning the emptying button or the cylinder
  • Large display ensures easy reading of the set volume value
  • Cones are indipendent, so the required force for securing the 8 or 12 tips is uniformly distributed
  • Minimum pressure required for inserting and ejecting tips
  • Movement of the cones allows the micropipette to oscillate during the loading operations
  • Lower part can freely turn 360°, the ejector is easily adjustable to house a large variety of tips and can be removed for cleaning
  • Fully autoclavable at +121 °C for 20 minutes, without having to recalibrate them

Certifications: EC/IVD-compliant, in accordance with EN IS 8655

Delivery information: Supplied with individual calibration certificates.

Specifications
  • Volume:
    50 - 300 µl
  • Accuracy:
    ±1,6 (50 µl); ±1,0 (300 µl) %
  • Imprecision:
    ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Stand for 3 pipettors, 145×70 mm
    Stand for 3 pipettors, 145×70 mm
    KART1315
    Transparent
    Log in for availability
    1 * 1 ST - 81,90 €