Multi-way stopcocks, Discofix®
Two-way stopcocks, Blue
Catalog # 720-2579
Supplier: B. Braun
Jump to:
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
- Continuous flow channels for accurate pressure monitoring
- Swivel lock can be moved axially or radially for fast and secure connection
- PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Specifications
- Colour:Blue
- Description:Two-way stopcocks