Wright's stain for microscopical staining
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Wright's stain for microscopical staining
Catalog # A17903.22
Supplier: Thermo Fisher Scientific
CAS Number: 68988-92-1
Jump to:
Product Details & Documents
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- CAS Number:68988-92-1
- MDL Number:MFCD00082143
- Density:0.8 g/cm³ (20 °C)
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



