Isosorbide dimethyl ether ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Isosorbide dimethyl ether ≥98%
Catalog # A17516.22
Supplier: Thermo Fisher Scientific
CAS Number: 5306-85-4
Springe zu:
Product Details & Documents
Purity: 98% CAS No.: 5306-85-4 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 108°(226°F) Refractive Index: 1.4620 Melting Point: -70° Boiling Point: 93-95°/0.1mm Beilstein Reference: 80854 MDL No.: MFCD00064828
Synonyme: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
- CAS Nummer:5306-85-4
- Formel:C₈H₁₄O₄
- MDL-Nummer:MFCD00064828
- Molekulargewicht:174.20 g/mol
- Siedepunkt:93…95 °C (0.1 mmHg)
- Schmelzpunkt:–70 °C
- Dichte:1.165
- Blitzpunkt:108 °C (226 °F)
- Lagertemperatur:Ambient
- EINECS:226-159-8
Documents
Spezifikationen
Spezifikationen
- Pack type:Glass bottle for solids
- Pk:100 g



