Order Entry
Austria
Orders LinkContactUsLinkComponent
Isosorbide dimethyl ether ≥98%

Isosorbide dimethyl ether ≥98%

Glass bottle for solids, 25 g
Catalog # A17516.14
Isosorbide dimethyl ether ≥98%
Catalog # A17516.14
CAS Number:  5306-85-4
Isosorbide dimethyl ether ≥98%
Isosorbide dimethyl ether ≥98%

Glass bottle for solids, 25 g

CAS Number:  5306-85-4
Price
61,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 5306-85-4 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 108°(226°F) Refractive Index: 1.4620 Melting Point: -70° Boiling Point: 93-95°/0.1mm Beilstein Reference: 80854 MDL No.: MFCD00064828

Synonyms:  (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan
  • CAS Number:5306-85-4
  • Formula:C₈H₁₄O₄
  • MDL Number:MFCD00064828
  • Molecular Weight:174.20 g/mol
  • Boiling Pt:93…95 °C (0.1 mmHg)
  • Melting Pt:–70 °C
  • Density:1.165
  • Flash Pt:108 °C (226 °F)
  • Storage Temperature:Ambient
  • EINECS:226-159-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.