Order Entry
Austria
Orders LinkContactUsLinkComponent
Iodoacetamide 98% stabilised

Iodoacetamide 98% stabilised

Glass bottle for solids, 25 g
Catalog # A14715.14
Toxic
Irritant
Iodoacetamide 98% stabilised
Catalog # A14715.14
CAS Number:  144-48-9
Iodoacetamide 98% stabilised
Iodoacetamide 98% stabilised

Glass bottle for solids, 25 g

CAS Number:  144-48-9
Price
98,30 €
Log in for availability
1 * 25 g
Product Details & Documents

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation.

2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation

Synonyms:  2-Iodoacetamide
  • CAS Number:144-48-9
  • Formula:ICH₂CONH₂
  • MDL Number:MFCD00008028
  • Molecular Weight:184.96 g/mol
  • Boiling Pt:266 °C (1013 hPa)
  • Melting Pt:92…95 °C (dried)
  • Storage Temperature:Refrigerator
  • EINECS:205-630-1
  • UN:2811
  • ADR:6.1,III
  •  :Moistened with water ∼5-8%
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications