Order Entry
Austria
Orders LinkContactUsLinkComponent
Cyclopentane 95%

Cyclopentane 95%

Glass bottle, 100 mL
Catalog # A14697.AE
Flammable
Cyclopentane 95%
Catalog # A14697.AE
CAS Number:  287-92-3
Cyclopentane 95%
Cyclopentane 95%

Glass bottle, 100 mL

CAS Number:  287-92-3
Price
40,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 95% CAS No.: 287-92-3 Molecular Formula: C5H10 Formula Weight: 70.14 Flash Point: -37°(-34°F) Physical Form: Liquid Density: 0.748 Refractive Index: 1.4060 Appearance: Colourless Odour: Petrol-like Melting Point: -95° Boiling Point: 47-49° Beilstein Reference: 1900195 Merck Reference: 14,2741 UN No.: 1146 MDL No.: MFCD00001356

Synonyms:  Pentamethylene
  • CAS Number:287-92-3
  • Formula:C₅H₁₀
  • MDL Number:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:14,2741
  • Boiling Pt:47…49 °C
  • Melting Pt:–95 °C
  • Density:0.748
  • Flash Pt:–37 °C (–34 °F)
  • Storage Temperature:Ambient
  • EINECS:206-016-6
  • UN:1146
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.