Orders LinkContactUsLinkComponent
Acetylacetone ≥99%
Glass bottle, 500 mL
  A14117.AP
Acetylacetone ≥99%
  A14117.AP
 :  123-54-6
Acetylacetone ≥99%
Acetylacetone ≥99%
Glass bottle, 500 mL
 :  123-54-6
48,40 €
1 * 500 mL
Purity: 99% CAS No.: 123-54-6 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 34°(93°F) Physical Form: Clear liquid Density: 0.973 Refractive Index: 1.4520 Appearance: Colourless Melting Point: -23° Boiling Point: 140-141° Beilstein Reference: 741937 Merck Reference: 14,81 UN No.: 2310 MDL No.: MFCD00008787
 :  ACAC, 2,4-Pentanedione
  •  :123-54-6
  •  :CH₃COCH₂COCH₃
  •  :MFCD00008787
  •  :100.12 g/mol
  •  :14,81
  •  :140…141 °C
  •  :–23 °C
  •  :0.973
  •  :34 °C (93 °F)
  •  :Ambient
  •  :204-634-0
  •  :2310
  •  :3,III
  • Pack type:
    Glass bottle
  • Pk:
    500 mL