Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetylacetone ≥99%

Acetylacetone ≥99%

Glass bottle, 2.500 mL
Catalog # A14117.0F
Toxic
Flammable
Acetylacetone ≥99%
Catalog # A14117.0F
CAS Number:  123-54-6
Acetylacetone ≥99%
Acetylacetone ≥99%

Glass bottle, 2.500 mL

CAS Number:  123-54-6
Price
173,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 123-54-6 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 34°(93°F) Physical Form: Clear liquid Density: 0.973 Refractive Index: 1.4520 Appearance: Colourless Melting Point: -23° Boiling Point: 140-141° Beilstein Reference: 741937 Merck Reference: 14,81 UN No.: 2310 MDL No.: MFCD00008787

Synonyms:  ACAC, 2,4-Pentanedione
  • CAS Number:123-54-6
  • Formula:CH₃COCH₂COCH₃
  • MDL Number:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Merck Index:14,81
  • Boiling Pt:140…141 °C
  • Melting Pt:–23 °C
  • Density:0.973
  • Flash Pt:34 °C (93 °F)
  • Storage Temperature:Ambient
  • EINECS:204-634-0
  • UN:2310
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.