Orders LinkContactUsLinkComponent
Sodium fluoride ≥99%
Sodium fluoride ≥99%
Glass bottle for solids, 5.000 g
  A13019.0I
Sodium fluoride ≥99%
  A13019.0I
 :  7681-49-4
Sodium fluoride ≥99%
Sodium fluoride ≥99%
Glass bottle for solids, 5.000 g
 :  7681-49-4
362,00 €
1 * 5.000 g
Purity: 99% CAS No.: 7681-49-4 Molecular Formula: FNa Formula Weight: 41.99 Refractive Index: 1.336 Odour: Odourless Boiling Point: 1704° Merck Reference: 14,8618 UN No.: 1690 MDL No.: MFCD00003524
  •  :7681-49-4
  •  :NaF
  •  :MFCD00003524
  •  :41.99 g/mol
  •  :14,8618
  •  :1704 °C
  •  :993 °C
  •  :2.78
  •  :Ambient
  •  :231-667-8
  •  :1690
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.