Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Halogen bulbs

Halogen bulb, 6 V, 30 W
  630-2579
 :   ZEISS
Halogen bulbs
  630-2579
 :  ZEISS
Halogen bulbs

Halogen bulb, 6 V, 30 W

 
 
   
1 * 1 ST
 
 
  • Description:
    Halogen bulb
  • Seq:
    911
  • Power:
    30 W
  • Voltage:
    6 V