Order Entry
Austria
Orders LinkContactUsLinkComponent
Phenyl chloroformate ≥99%

Phenyl chloroformate ≥99%

Glass bottle for solids, 250 g
Catalog # A12618.30
Toxic
Corrosive
Irritant
Phenyl chloroformate ≥99%
Catalog # A12618.30
CAS Number:  1885-14-9
Phenyl chloroformate ≥99%
Phenyl chloroformate ≥99%

Glass bottle for solids, 250 g

CAS Number:  1885-14-9
Price
87,50 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 1885-14-9 Molecular Formula: ClCO2C6H5 Formula Weight: 156.57 Storage Temperature: 2°C to 8°C Flash Point: 76°(169°F) Refractive Index: 1.5110 Melting Point: -38° Boiling Point: 188-189° Beilstein Reference: 606778 UN No.: 2746 MDL No.: MFCD00000637

  • CAS Number:1885-14-9
  • Formula:C₇H₅ClO₂
  • MDL Number:MFCD00000637
  • Molecular Weight:156.57 g/mol
  • Boiling Pt:188…189 °C
  • Melting Pt:–38 °C
  • Density:1.246
  • Flash Pt:76 °C (169 °F)
  • EINECS:217-547-8
  • UN:2746
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.