Order Entry
Austria
Orders LinkContactUsLinkComponent
Fumaric acid ≥99%

Fumaric acid ≥99%

Glass bottle for solids, 2.500 g
Catalog # A10976.0E
Irritant
Fumaric acid ≥99%
Catalog # A10976.0E
CAS Number:  110-17-8
Fumaric acid ≥99%
Fumaric acid ≥99%

Glass bottle for solids, 2.500 g

CAS Number:  110-17-8
Price
70,30 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 110-17-8 Molecular Formula: HO2CCH=CHCO2H Formula Weight: 116.07 Flash Point: 273°(523°F) Odour: Odourless pH: 2.1 Melting Point: ca 299° subl. Beilstein Reference: 605763 Merck Reference: 14,4287 MDL No.: MFCD00002700

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:14,4287
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:∼ 299 °C (subl.)
  • Density:1.625
  • Flash Pt:273 °C (523 °F)
  • Storage Temperature:Ambient
  • EINECS:203-743-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications