Orders LinkContactUsLinkComponent
Fumaric acid ≥99%
Fumaric acid ≥99%
Glass bottle for solids, 10.000 g
  A10976.0C
Fumaric acid ≥99%
  A10976.0C
 :  110-17-8
Fumaric acid ≥99%
Fumaric acid ≥99%
Glass bottle for solids, 10.000 g
 :  110-17-8
220,00 €
1 * 10.000 g
Purity: 99% CAS No.: 110-17-8 Molecular Formula: HO2CCH=CHCO2H Formula Weight: 116.07 Flash Point: 273°(523°F) Odour: Odourless pH: 2.1 Melting Point: ca 299° subl. Beilstein Reference: 605763 Merck Reference: 14,4287 MDL No.: MFCD00002700
 :  trans-1,2-Ethylentricarboxylic acid
  •  :110-17-8
  •  :C₄H₄O₄
  •  :MFCD00002700
  •  :116.07 g/mol
  •  :14,4287
  •  :356 °C (1013 hPa)
  •  :∼ 299 °C (subl.)
  •  :1.625
  •  :273 °C (523 °F)
  •  :Ambient
  •  :203-743-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g