Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer HF-Improved

Buffer HF-Improved

Buffer HF-Improved, 1 L
Catalog # 44627.K2
Buffer HF-Improved
Catalog # 44627.K2
CAS Number:  
Buffer HF-Improved
Buffer HF-Improved

Buffer HF-Improved, 1 L

CAS Number:  
Price
118,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Buffer used for pretreatment of planar silicon devices when plating with nickel plating solution products 44069 and 44070.

Density: 1.12
Appearance: Colorless
Odor: Acrid
Boiling Point: 103…105°
UN No.: 2817

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Buffer HF-Improved
  • UN No.:
    2817
  • Cat. No.:
    44627.K2
  • Odour/scent:
    Acrid
  • MDL No.:
    MFCD00132734
  • Density:
    1.12 g/mL
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.