Order Entry
Austria
Orders LinkContactUsLinkComponent
Transition metals, plasma standard solution, Specpure®

Transition metals, plasma standard solution, Specpure®

Transition metals, plasma standard solution, 5% HNO₃, 500 mL
Catalog # 44518.AP
Transition metals, plasma standard solution, Specpure®
Catalog # 44518.AP
CAS Number:  
Transition metals, plasma standard solution, Specpure®
Transition metals, plasma standard solution, Specpure®

Transition metals, plasma standard solution, 5% HNO₃, 500 mL

CAS Number:  
Price
601,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 µg/ml

UN No.: UN3264
MDL No.: MFCD00151266

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    Transition metals, plasma standard solution
  • Cat. No.:
    44518.AP
  • MDL No.:
    MFCD00151266

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.