Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH buffer solutions, Specpure®

4,00, ±0,01, Phthalate, Red, 25 °C, Plastic container, 20 L
Catalog # 38713.M4
pH buffer solutions, Specpure®
Catalog # 38713.M4
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

4,00, ±0,01, Phthalate, Red, 25 °C, Plastic container, 20 L

CAS Number:  
Price
384,00 €
Log in for availability
1 * 20 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic container
  • pH buffer type:
    Phthalate
  • Pk:
    20 L
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,01
  • pH value:
    4,00

Specifications