ContactUsLinkComponent
Flask volumetric, capacity 25ml, class A, amber graduated, polystoppered, tolerance 0.04ml, height 110mm, NS-ground 10/19, draduatio n color white, conformity certified. 1 * 1 items
Catalog # HIRS264017037
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 25ml, class A, amber graduated, polystoppered, tolerance 0.04ml, height 110mm, NS-ground 10/19, draduatio n color white, conformity certified. 1 * 1 items
Catalog # HIRS264017037
Supplier:  HIRSCHMANN
Frequently Bought Together