Orders LinkContactUsLinkComponent
Mailing boxes
Mailing boxes
Mailing boxes with enclosed foam bag for 1 - 10 containers, 220×170×40 mm
Catalog # SARS95.902
Marketsource
Supplier:   SARSTEDT
Mailing boxes
Catalog # SARS95.902
Supplier:  SARSTEDT
Marketsource
Mailing boxes
Mailing boxes with enclosed foam bag for 1 - 10 containers, 220×170×40 mm
Price
396,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
  • With printed label
  • Lightweight
  • Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Specifications
  • Description:
    Mailing boxes with enclosed foam bag for 1 - 10 containers
  • Seq:
    03
  • W×D×H:
    220×170×40 mm