Orders LinkContactUsLinkComponent
Microtubes, with Thread
Micro tubes without screw cap, without skirted base, 0,5 ml, 5000/case, 0,5 ml
Artikelnr. SARS72.733.010
Marketsource
Anbieter:   SARSTEDT
Microtubes, with Thread
Artikelnr. SARS72.733.010
Anbieter:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes without screw cap, without skirted base, 0,5 ml, 5000/case, 0,5 ml
Preis
292,00 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
  • Packed:
    5000/case
  • Recommended working volume:
    0,5 ml
  • Capacity:
    0,5 ml
  • Description:
    Micro tubes without screw cap, without skirted base
  • Seq:
    9