Orders LinkContactUsLinkComponent
Microtubes, with Thread
Micro tubes without screw cap, skirted base, 1,5 ml, 5000/case
Catalog # SARS72.703
Marketsource
Supplier:   SARSTEDT
Microtubes, with Thread
Catalog # SARS72.703
Supplier:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes without screw cap, skirted base, 1,5 ml, 5000/case
Price
210,00 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
  • Packed:
    5000/case
  • Capacity:
    1,5 ml
  • Description:
    Micro tubes without screw cap, skirted base
  • Seq:
    10