Microtubes
Tubes with screw cap assembled, sterile, with writing space and graduation, 2,0 ml, 5000/case, 2,0 ml
Catalog # SARS72.694.107
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent
- Excellent chemical resistance
- Excellent temperature resistance
Without or with skirted base.
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:5000/case
- Recommended working volume:2,0 ml
- Capacity:2,0 ml
- Description:Tubes with screw cap assembled, sterile, with writing space and graduation
Accessories
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, blue
Product Information
- Catalog #AvailabilityPrice
- Screw cap with seperate retaining loop, neutralScrew cap with seperate retaining loop, neutralSARS65.712Log in for availability1 * 5.000 ST - 218,00 €
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, neutral
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, red
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, violet
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, white
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, black
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, yellow
Product Information
- Catalog #AvailabilityPrice
- Colour coded inserts, green