Orders LinkContactUsLinkComponent
Microtubes, with Thread
Microtubes, with Thread
Micro tubes with skirted base, without screw cap, without knurls, 2,0 ml, 5000/case
Catalog # SARS72.664
Marketsource
Supplier:   SARSTEDT
Microtubes, with Thread
Catalog # SARS72.664
Supplier:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes with skirted base, without screw cap, without knurls, 2,0 ml, 5000/case
Price
166,00 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Documents
Specifications
  • Packed:
    5000/case
  • Capacity:
    2,0 ml
  • Description:
    Micro tubes with skirted base, without screw cap, without knurls
  • Seq:
    15