Microtubes, Prepared
Microtubes, prepared, plasma-lithium heparin, with screw cap closure, 1,3 ml, 1000/case
Catalog # SARS41.1393.005
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent with PP screw cap.
- Type of preparation: Trisodium citrate solution
- With paper label
- Graduated
- Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specifications
- Packed:1000/case
- Capacity:1,3 ml
- Description:Microtubes, prepared, plasma-lithium heparin, with screw cap closure
- Seq:5